missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tellurium powder, -30 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01107414

 View more versions of this product

Catalog No. AA1107414


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tellurium powder
(metals basis)
MFCD00134062
25 g
PORWMNRCUJJQNO-UHFFFAOYSA-N
tellurium
6327182
99.99%
Odorless
13494-80-9
Te
UN3288
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
[Te]
127.60
CHEBI:30452
−30 Mesh
Powder

Chemical Identifiers

13494-80-9
127.60
PORWMNRCUJJQNO-UHFFFAOYSA-N
6327182
tellurium
Te
MFCD00134062
tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
CHEBI:30452
[Te]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 236-813-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only