missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TEMED, Approx. 99%, Molecular Biology Reagent, MP Biomedicals™
N,N,N',N'-Tetramethylethylenediamine is used to catalyze the formation of free radicals from ammonium persulfate or riboflavin
Supplier: MP Biomedicals Inc 0219401980
Specifications
N,N,N',N'-Tetramethylethylenediamine | |
110-18-9 | |
Approx. 99% | |
MFCD00008335 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
CN(C)CCN(C)C | |
116.208 | |
CHEBI:32850 | |
Molecular Biology Reagent |
FTIR | |
0.775g/mL at 20°C | |
C6H16N2 | |
100 mL | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
N,N,N',N'-tetramethylethane-1,2-diamine | |
8037 | |
116.2 | |
Liquid |
Chemical Identifiers
110-18-9 | |
116.208 | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
8037 | |
N,N,N',N'-tetramethylethane-1,2-diamine |
C6H16N2 | |
MFCD00008335 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
CHEBI:32850 | |
CN(C)CCN(C)C |
Safety and Handling
EINECSNumber : 203-744-6