missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TEMED (Electrophoresis), Fisher BioReagents
For polymerization of acrylamide/bis-acrylamide
Supplier: Fisher BioReagents BP150100
Description
TEMED is a commonly used catalyst for the polymerization of acrylamide/bis-acrylamide.Specifications
TEMED | |
-55°C | |
0.770g/cm³ | |
120°C | |
≥97 % (by titration) | |
C6H16N2 | |
130 mL | |
15, 9277 | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
N,N,N',N'-tetramethylethane-1,2-diamine | |
4.00g/mol | |
CHEBI:32850 | |
≥97% | |
Liquid |
110-18-9 | |
Colorless | |
8.0 to 8.5 | |
DNase-, RNase- and Protease-Free | |
Amber Glass | |
(CH3)2NCH2CH2N(CH3)2 | |
2372 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
CN(C)CCN(C)C | |
116.208 | |
8037 | |
116.21 | |
Electrophoresis |
Chemical Identifiers
110-18-9 | |
116.208 | |
temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl | |
CHEBI:32850 | |
CN(C)CCN(C)C |
C6H16N2 | |
KWYHDKDOAIKMQN-UHFFFAOYSA-N | |
8037 | |
N,N,N',N'-tetramethylethane-1,2-diamine |
Safety and Handling
DOTInformation : DOT Class 3, : Flammable Liquid
Recommended Storage : RT