Tempshield™ Cryo-Apron™
Protection from splashes and contact cold in cryogenic and ultra-cold environments to -196°C.
$500.50 - $568.30
Catalog Number | Mfr. No. | Length (Metric) | Length (English) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Length (English) | Price | Quantity | ||||
17-540A
|
Tempshield™
CA36 |
91.4 cm | 36 in. |
Each for $518.06
Case of 10 Each for $4,474.30
|
|
||||
17-540C
|
Tempshield™
CA48 |
122 cm | 48 in. |
Each for $500.50
Case of 10 Each for $4,300.20
|
|
||||
17-540B
|
Tempshield™
CA42 |
106 cm | 42 in. |
Each for $568.30
Case of 10 Each for $4,908.10
|
|
||||
Description
Protection from splashes and contact cold in cryogenic and ultra-cold environments to -196°C.
Protection from splashes and contact cold in cryogenic and ultra-cold environments to -196°C.
- Waterproof, lightweight
- Adjustable straps at neck and waist
- All aprons are 24 in. (61cm) wide
- Color: blue
Specifications
Blue |