Tempshield™ Cryo-Gloves™ Blue Wrist Length Cryogenic Gloves DFS Item
State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
$407.58 - $448.53
Specifications
Certifications/Compliance (Europe) EN 420, EN 511
Description Wrist Length
Color Blue
Hand Specific
Product Type Cryogenic Gloves
View More Specs
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-276533 DFS Item
View Documents
Tempshield™
WRS
Small
Pair for $444.91
Case of 10 Pair for $4,050.70
19-276535 DFS Item
View Documents
Tempshield™
WRL
Large
Pair for $448.53
Case of 10 Pair for $4,083.70
19-276534 DFS Item
View Documents
Tempshield™
WRM
Medium
Pair for $407.58
Case of 10 Pair for $3,710.90
Description
Description
State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
Specifications
EN 420, EN 511
Blue
Cryogenic Gloves
Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
-320°F
11.3 to 12.5 in.
Wrist Length
Specific
Yes
Cryo-Gloves™
-196°C
29 to 32.5 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'