missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Terbium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals
$133.48 - $480.29
Chemical Identifiers
CAS | 14284-95-8 |
---|---|
Molecular Formula | C15H21O6Tb |
Molecular Weight (g/mol) | 456.25 |
MDL Number | MFCD00013503 |
InChI Key | MAPWZFZLNRMAMO-UHFFFAOYSA-N |
Synonym | terbium iii acetylacetonate, terbium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 |
PubChem CID | 129878174 |
IUPAC Name | (Z)-4-hydroxypent-3-en-2-one;terbium |
SMILES | [Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Description
Terbium(III) 2,4-pentanedionate is used as an active pharmaceutical ingredient. It is also used as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14284-95-8 | |
456.25 | |
MAPWZFZLNRMAMO-UHFFFAOYSA-N | |
129878174 | |
[Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C15H21O6Tb | |
MFCD00013503 | |
terbium iii acetylacetonate, terbium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 | |
(Z)-4-hydroxypent-3-en-2-one;terbium |
Specifications
14284-95-8 | |
2 g | |
MFCD00013503 | |
Insoluble in water. | |
[Tb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
456.25 | |
456.25 | |
Odorless | |
(REO) |
Powder | |
C15H21O6Tb | |
terbium iii acetylacetonate, terbium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 | |
MAPWZFZLNRMAMO-UHFFFAOYSA-N | |
(Z)-4-hydroxypent-3-en-2-one;terbium | |
129878174 | |
99.9% | |
Terbium(III) 2,4-pentanedionate |
Safety and Handling
EINECSNumber : 238-195-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only