missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Terbium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

$87.44 - $1438.08

Chemical Identifiers

CAS 13842-67-6
MDL Number MFCD00149872
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1459604
View Documents
Thermo Scientific Chemicals
01459604
2 g
Each for $87.44
 
AA1459609
View Documents
Thermo Scientific Chemicals
01459609
10 g
Each for $266.62
 
AA1459618
View Documents
Thermo Scientific Chemicals
01459618
50 g
Each for $1,438.08
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13842-67-6
MFCD00149872

Specifications

360°C (in -8H2O)
Crystalline
MFCD00149872
99.9%
Terbium(III) sulfate octahydrate
13842-67-6
2 g
750.19 (606.05 Anhydrous)
Reacton™
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-213-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only