missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Terbium sputtering target, 50.8mm dia x 3.18mm thick, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 45738KS
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Terbium sputtering target | |
7440-27-9 | |
MFCD00011256 | |
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic | |
GZCRRIHWUXGPOV-UHFFFAOYSA-N | |
terbium | |
23958 |
50.8mm dia. x 3.18mm thick | |
Tb | |
1 Ea. | |
Insoluble in water. | |
[Tb] | |
158.93 | |
CHEBI:33376 |
Chemical Identifiers
7440-27-9 | |
158.93 | |
GZCRRIHWUXGPOV-UHFFFAOYSA-N | |
23958 | |
terbium |
Tb | |
MFCD00011256 | |
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic | |
CHEBI:33376 | |
[Tb] |
Safety and Handling
EINECSNumber : 231-137-6
TSCA : Yes
Recommended Storage : Ambient temperatures