Learn More
Thermo Scientific Chemicals Tergitol(TM) 15-S-9
$82.31 - $144.74
Chemical Identifiers
CAS | 68131-40-8 |
---|---|
Molecular Formula | [C2H4O]n C13H27O |
MDL Number | MFCD00132411 |
SMILES | CCCCCCCC(CCCCC)OCCO-* |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC464252500
|
Thermo Scientific Chemicals
464252500 |
250 mL |
Each for $82.31
|
|
||||||
AC464250010
|
Thermo Scientific Chemicals
464250010 |
1 L |
Each for $144.74
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68131-40-8 | |
[C2H4O]n C13H27O | |
MFCD00132411 | |
CCCCCCCC(CCCCC)OCCO-* |
Specifications
68131-40-8 | |
250 mL | |
1.01 | |
MFCD00132411 | |
CCCCCCCC(CCCCC)OCCO-* |
1.0100 g/mL | |
193°C | |
[C2H4O]n C13H27O | |
Solubility aq. soln.: soluble. | |
Tergitol* 15-S-9 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
RUO – Research Use Only