missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butanol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 390690025

 View more versions of this product

Catalog No. AC390690025

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butanol
For analysis ACS
0.7800g/mL
2.5 L
Authentic
Liquid
0.001% max.
0.0005 meq/g max.
(CH3)3COH
3.35 mPa.s (30°C)
0.1% max. (K.F.)
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
DKGAVHZHDRPRBM-UHFFFAOYSA-N
2-methylpropan-2-ol
6386
74.12
ACS Reagent
25.0°C to 25.5°C
75-65-0
83.0°C
11°C
Plastic Bottle
1.3860 to 1.3880
0.78
C4H10O
MFCD00004464
1120
15, 1544
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, acetone
CC(C)(C)O
74.123
CHEBI:45895
≥99.5%

Chemical Identifiers

75-65-0
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
2-methylpropan-2-ol
C4H10O
MFCD00004464
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CHEBI:45895
CC(C)(C)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only