Learn More
tert-Butanol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 390690010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
tert-Butanol | |
For analysis ACS | |
0.7800g/mL | |
1 L | |
Authentic | |
Liquid | |
0.001% max. | |
0.0005 meq/g max. | |
(CH3)3COH | |
3.35 mPa.s (30°C) | |
0.1% max. (K.F.) | |
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol | |
DKGAVHZHDRPRBM-UHFFFAOYSA-N | |
2-methylpropan-2-ol | |
6386 | |
74.12 | |
ACS Reagent |
25.0°C to 25.5°C | |
75-65-0 | |
83.0°C | |
11°C | |
Plastic Bottle | |
1.3860 to 1.3880 | |
0.78 | |
C4H10O | |
MFCD00004464 | |
1120 | |
15, 1544 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, acetone | |
CC(C)(C)O | |
74.123 | |
CHEBI:45895 | |
≥99.5% |
Chemical Identifiers
75-65-0 | |
74.123 | |
DKGAVHZHDRPRBM-UHFFFAOYSA-N | |
6386 | |
2-methylpropan-2-ol |
C4H10O | |
MFCD00004464 | |
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol | |
CHEBI:45895 | |
CC(C)(C)O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only