missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-butyl 4-(4-formylbenzyloxy)piperidine-1-carboxylate, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC67904DA

 View more versions of this product

Catalog No. CC67904DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Tert-butyl 4-(4-formylbenzyloxy)piperidine-1-carboxylate
89.5
90%
C18H25NO4
1g
WQFHJESQGSVMDW-UHFFFAOYSA-N
tert-butyl 4-[(4-formylphenyl)methoxy]piperidine-1-carboxylate
33589526
90%
1080028-74-5
100.0
Amber Glass Bottle
MFCD11841070
tert-butyl 4-4-formylbenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-formylphenyl methoxy piperidine-1-carboxylate, 1-4-formylbenzyl oxy piperidine, n-boc protected, tert-butyl 4-4-formylbenzyl oxy piperidine-1-carboxylate, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy methyl benzaldehyde
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)C=O
319.401
319.4

Chemical Identifiers

1080028-74-5
319.401
WQFHJESQGSVMDW-UHFFFAOYSA-N
33589526
CC(C)(C)OC(=O)N1CCC(CC1)OCC2=CC=C(C=C2)C=O
C18H25NO4
MFCD11841070
tert-butyl 4-4-formylbenzyloxy piperidine-1-carboxylate, tert-butyl 4-4-formylphenyl methoxy piperidine-1-carboxylate, 1-4-formylbenzyl oxy piperidine, n-boc protected, tert-butyl 4-4-formylbenzyl oxy piperidine-1-carboxylate, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy methyl benzaldehyde
tert-butyl 4-[(4-formylphenyl)methoxy]piperidine-1-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions