missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate, ≥90%, Thermo Scientific™

Chemical Identifiers

CAS 681508-91-8
Molecular Formula C17H27N3O2
Molecular Weight (g/mol) 305.422
MDL Number MFCD08060503
InChI Key VERAVZPFPZKVCT-UHFFFAOYSA-N
Synonym tert-butyl 4-4-methylamino methyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-methylaminomethyl phenyl piperazine-1-carboxylate
PubChem CID 7164564
IUPAC Name tert-butyl 4-[4-(methylaminomethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CNC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC30046CB
SDS View Documents
Thermo Scientific Chemicals
CC30046CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30046DA
SDS View Documents
Thermo Scientific Chemicals
CC30046DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

681508-91-8
305.422
VERAVZPFPZKVCT-UHFFFAOYSA-N
7164564
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CNC
C17H27N3O2
MFCD08060503
tert-butyl 4-4-methylamino methyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-methylaminomethyl phenyl piperazine-1-carboxylate
tert-butyl 4-[4-(methylaminomethyl)phenyl]piperazine-1-carboxylate

Specifications

681508-91-8
100.0
Amber Glass Bottle
MFCD08060503
tert-butyl 4-4-methylamino methyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-methylaminomethyl phenyl piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CNC
305.422
305.42
Tert-butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate
89.5
90%
C17H27N3O2
250mg
VERAVZPFPZKVCT-UHFFFAOYSA-N
tert-butyl 4-[4-(methylaminomethyl)phenyl]piperazine-1-carboxylate
7164564
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'