Learn More
tert-Butyl 4-aminobutyrate hydrochloride, 97%, Thermo Scientific Chemicals
$109.58 - $438.29
Chemical Identifiers
CAS | 58640-01-0 |
---|---|
Molecular Formula | C8H18ClNO2 |
Molecular Weight (g/mol) | 195.687 |
MDL Number | MFCD00270592 |
InChI Key | CZVNLCOJFFYZPG-UHFFFAOYSA-N |
Synonym | tert-butyl 4-aminobutanoate hydrochloride, h-gamma-abu-otbu hydrochloride, h-gamma-abu-otbu hcl, h-gamma-abu-otbu hcl, gamma-aminobutyric acid t-butyl ester hcl, 4-aminobutyric acid tert-butyl ester hydrochloride, 4-aminobutyric acid tert-butyl ester hydrochloride, a-abu-otbu.hcl, h-gaba-otbu . hcl, h-gaba-otbu . hcl, h-.gamma.-abu-otbu.hcl |
PubChem CID | 24750478 |
IUPAC Name | tert-butyl 4-aminobutanoate;hydrochloride |
SMILES | CC(C)(C)OC(=O)CCCN.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6593603
|
Thermo Scientific Chemicals
H6593603 |
1 g |
Each for $109.58
|
|
||||||
AAH6593606
|
Thermo Scientific Chemicals
H6593606 |
5 g |
Each for $438.29
|
|
||||||
Description
Employed as an important intermediate for raw material for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as an important intermediate for raw material for pharmaceutical.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
58640-01-0 | |
195.687 | |
CZVNLCOJFFYZPG-UHFFFAOYSA-N | |
24750478 | |
CC(C)(C)OC(=O)CCCN.Cl |
C8H18ClNO2 | |
MFCD00270592 | |
tert-butyl 4-aminobutanoate hydrochloride, h-gamma-abu-otbu hydrochloride, h-gamma-abu-otbu hcl, h-gamma-abu-otbu hcl, gamma-aminobutyric acid t-butyl ester hcl, 4-aminobutyric acid tert-butyl ester hydrochloride, 4-aminobutyric acid tert-butyl ester hydrochloride, a-abu-otbu.hcl, h-gaba-otbu . hcl, h-gaba-otbu . hcl, h-.gamma.-abu-otbu.hcl | |
tert-butyl 4-aminobutanoate;hydrochloride |
Specifications
58640-01-0 | |
C8H18ClNO2 | |
MFCD00270592 | |
CZVNLCOJFFYZPG-UHFFFAOYSA-N | |
tert-butyl 4-aminobutanoate;hydrochloride | |
195.687 | |
195.69 | |
Powder |
White | |
H2NCH2CH2CH2CO2C(CH3)3·HCl | |
tert-butyl 4-aminobutanoate hydrochloride, h-gamma-abu-otbu hydrochloride, h-gamma-abu-otbu.hcl, gamma-aminobutyric acid t-butyl ester hcl, gamma-aminobutyric acid t-butyl ester hcl, 4-aminobutyric acid tert-butyl ester hydrochloride, 4-aminobutyric acid tert-butyl ester hydrochloride, a-abu-otbu.hcl, h-gaba-otbu . hcl, h-gaba-otbu . hcl, h-.gamma.-abu-otbu.hcl | |
CC(C)(C)OC(=O)CCCN.Cl | |
1 g | |
24750478 | |
97% | |
tert-Butyl 4-aminobutyrate hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only