Learn More
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
$68.66 - $451.07
Chemical Identifiers
CAS | 540-88-5 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00008807 |
InChI Key | WMOVHXAZOJBABW-UHFFFAOYSA-N |
Synonym | t-butyl acetate, acetic acid tert-butyl ester, tert-butyl ethanoate, acetic acid, 1,1-dimethylethyl ester, texaco lead appreciator, acetic acid, tert-butyl ester, tert-butylacetate, tert butyl acetate, 1,1-dimethylethyl acetate, 1,1-dimethyl acetate |
PubChem CID | 10908 |
IUPAC Name | tert-butyl acetate |
SMILES | CC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0885518
|
Thermo Scientific Chemicals
L0885518 |
50 g |
Each for $68.66
|
|
||||||
AAL0885530
|
Thermo Scientific Chemicals
L0885530 |
250 g |
Each for $154.85
|
|
||||||
AAL088550B
|
Thermo Scientific Chemicals
L088550B |
1000 g |
Each for $451.07
|
|
||||||
Description
tert-Butyl acetate is used as a solvent in the production of lacquers, enamels, inks, adhesives, thinners, industrial cleaners and coating blends It is also used as a process solvent for chemical reactions and polymerization. In synthetic organic chemistry, it is used as a tert-butyl group. It serves as an intermediate in the production of pharmaceuticals and synthetic flavorings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl acetate is used as a solvent in the production of lacquers, enamels, inks, adhesives, thinners, industrial cleaners and coating blends It is also used as a process solvent for chemical reactions and polymerization. In synthetic organic chemistry, it is used as a tert-butyl group. It serves as an intermediate in the production of pharmaceuticals and synthetic flavorings.
Solubility
Miscible with ether and ethanol. Immiscible with water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
540-88-5 | |
116.16 | |
WMOVHXAZOJBABW-UHFFFAOYSA-N | |
10908 | |
CC(=O)OC(C)(C)C |
C6H12O2 | |
MFCD00008807 | |
t-butyl acetate, acetic acid tert-butyl ester, tert-butyl ethanoate, acetic acid, 1,1-dimethylethyl ester, texaco lead appreciator, acetic acid, tert-butyl ester, tert-butylacetate, tert butyl acetate, 1,1-dimethylethyl acetate, 1,1-dimethyl acetate | |
tert-butyl acetate |
Specifications
540-88-5 | |
0.863 | |
4°C (39°F) | |
1.3865 | |
MFCD00008807 | |
UN1123 | |
14,1537 | |
Miscible with ether and ethanol. Immiscible with water. | |
CC(=O)OC(C)(C)C | |
116.16 | |
116.16 | |
tert-Butyl acetate |
-62°C | |
94°C to 96°C | |
C6H12O2 | |
CH3CO2C(CH3)3 | |
50 g | |
1699506 | |
t-butyl acetate, acetic acid tert-butyl ester, tert-butyl ethanoate, acetic acid, 1,1-dimethylethyl ester, texaco lead appreciator, acetic acid, tert-butyl ester, tert-butylacetate, tert butyl acetate, 1,1-dimethylethyl acetate, 1,1-dimethyl acetate | |
WMOVHXAZOJBABW-UHFFFAOYSA-N | |
tert-butyl acetate | |
10908 | |
99% |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H225-H332-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL ACETATES
EINECSNumber : 208-760-7
RTECSNumber : AF7400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only