missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl acetoacetate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1111536

 View more versions of this product

Catalog No. AAA1111536


Explore available promotions

Description

Description

tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ?,β - unsaturated ketones synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ∞,β - unsaturated ketones synthesis.

Solubility
Soluble in water (9g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

tert-Butyl acetoacetate
-38°C
5.7
76°C (169°F)
C8H14O3
MFCD00008811
1680303
Soluble in water (9g/L).
CC(=O)CC(=O)OC(C)(C)C
158.197
158.2
1694-31-1
0.961
71°C to 72°C (11 mmHg)
Pleasant
1.42
500 g
tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
JKUYRAMKJLMYLO-UHFFFAOYSA-N
tert-butyl 3-oxobutanoate
15538
≥98%

Chemical Identifiers

1694-31-1
158.197
JKUYRAMKJLMYLO-UHFFFAOYSA-N
15538
CC(=O)CC(=O)OC(C)(C)C
C8H14O3
MFCD00008811
tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
tert-butyl 3-oxobutanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 216-904-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only