missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Alcohol 99.0+%, TCI America™

$41.02 - $61.51

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B070625ML
View Documents
TCI America
B070625ML
25 mL
Each for $41.02
Only null left
 
B0706500ML
View Documents
TCI America
B0706500ML
500 mL
Each for $61.51
Only null left
 
Description

Description

Specifications

Chemical Identifiers

75-65-0
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
2-methylpropan-2-ol
C4H10O
MFCD00004464
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CHEBI:45895
CC(C)(C)O

Specifications

White
82°C
MFCD00004464
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CC(C)(C)O
74.123
CHEBI:45895
≥99.0% (GC)
Crystal-Lump at 20°C
75-65-0
C4H10O
1120
DKGAVHZHDRPRBM-UHFFFAOYSA-N
2-methylpropan-2-ol
6386
74.12
25 mL
tert-Butyl Alcohol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3049

RTECSNumber : EO1925000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'