missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl chloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A13004AU

 View more versions of this product

Catalog No. AAA13004AU


Explore available promotions

Description

Description

tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

tert-Butyl chloride
-26°C
50°C to 52°C
Mild
1.3848
1000 mL
1730872
tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
NBRKLOOSMBRFMH-UHFFFAOYSA-N
2-chloro-2-methylpropane
10486
≥98%
507-20-0
0.846
−27°C (−16°F)
C4H9Cl
MFCD00000816
UN1127
14,1562
Miscible with alcohol,benzene,chloroform and ether. Slightly miscible with water.
CC(C)(C)Cl
92.566
92.57

Chemical Identifiers

507-20-0
92.566
NBRKLOOSMBRFMH-UHFFFAOYSA-N
10486
CC(C)(C)Cl
C4H9Cl
MFCD00000816
tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
2-chloro-2-methylpropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES

EINECSNumber : 208-066-4

RTECSNumber : TX5040000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only