Learn More
tert-Butyl chloroacetate, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 107-59-5 |
---|---|
Molecular Formula | C6H11ClO2 |
Molecular Weight (g/mol) | 150 |
MDL Number | MFCD00000930 |
InChI Key | KUYMVWXKHQSIAS-UHFFFAOYSA-N |
Synonym | tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate |
PubChem CID | 66052 |
IUPAC Name | tert-butyl 2-chloroacetate |
SMILES | CC(C)(C)OC(=O)CCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC347230250
|
Thermo Scientific Chemicals
347230250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC347231000
|
Thermo Scientific Chemicals
347231000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC347235000
|
Thermo Scientific Chemicals
347235000 |
500g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
107-59-5 | |
150 | |
KUYMVWXKHQSIAS-UHFFFAOYSA-N | |
66052 | |
CC(C)(C)OC(=O)CCl |
C6H11ClO2 | |
MFCD00000930 | |
tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate | |
tert-butyl 2-chloroacetate |
Specifications
107-59-5 | |
48.0°C to 49.0°C (11.0mmHg) | |
Authentic | |
Glass bottle | |
1.4220 to 1.4240 | |
MFCD00000930 | |
02, III, 444 | |
1.053 | |
tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate | |
KUYMVWXKHQSIAS-UHFFFAOYSA-N | |
tert-butyl 2-chloroacetate | |
66052 | |
97% |
1.0530g/mL | |
46°C | |
96% min. (GC) | |
C6H11ClO2 | |
ClCH2CO2C(CH3)3 | |
25g | |
01,86; 03,36 | |
15, 1564 | |
Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in isopropanol,1,2-dichloroethane,toluene | |
CC(C)(C)OC(=O)CCl | |
150 | |
150 | |
tert-Butyl chloroacetate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Very toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Toxic if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 203-506-1
RUO â Research Use Only