missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl diethylphosphonoacetate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 397750050

 View more versions of this product

Catalog No. AC397750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
Specifications

Specifications

tert-Butyl diethylphosphonoacetate
1.0740g/mL
Authentic
Glass bottle
MFCD00075414
1.074
NFEGNISFSSLEGU-UHFFFAOYSA-N
tert-butyl 2-diethoxyphosphorylacetate
2773685
95%
27784-76-5
>110°C
94% min. (GC)
C10H21O5P
5g
tert-butyl diethylphosphonoacetate, tert-butyl 2-diethoxyphosphoryl acetate, diethyl boc-methyl phosphonate, diethoxy-phosphoryl-acetic acid tert-butyl ester, t-butyl diethylphosphonoacetate, diethyl phosphonoacetic acid tert-butyl ester, t-butyl diethyl phosphonoacetate, diethylphosphonoacetic acid tert-butyl ester, diethyl 2-tert-butoxycarbonylmethyl phosphonate, acetic acid, diethoxyphosphinyl-, 1,1-dimethylethyl ester
CCOP(=O)(CC(=O)OC(C)(C)C)OCC
50.23
50.23

Chemical Identifiers

27784-76-5
50.23
NFEGNISFSSLEGU-UHFFFAOYSA-N
2773685
CCOP(=O)(CC(=O)OC(C)(C)C)OCC
C10H21O5P
MFCD00075414
tert-butyl diethylphosphonoacetate, tert-butyl 2-diethoxyphosphoryl acetate, diethyl boc-methyl phosphonate, diethoxy-phosphoryl-acetic acid tert-butyl ester, t-butyl diethylphosphonoacetate, diethyl phosphonoacetic acid tert-butyl ester, t-butyl diethyl phosphonoacetate, diethylphosphonoacetic acid tert-butyl ester, diethyl 2-tert-butoxycarbonylmethyl phosphonate, acetic acid, diethoxyphosphinyl-, 1,1-dimethylethyl ester
tert-butyl 2-diethoxyphosphorylacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only