missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl ethyl malonate, 95%

CAS: 32864-38-3 | C9H16O4 | 188.223 g/mol

$300.10 - $910.90

Chemical Identifiers

CAS 32864-38-3
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00009193
InChI Key OCOBFMZGRJOSOU-UHFFFAOYSA-N
Synonym tert-butyl ethyl malonate, t-butyl ethyl malonate, propanedioic acid, 1,1-dimethylethyl ethyl ester, 1-tert-butyl 3-ethyl propanedioate, tert-butyl ethyl propanedioate, malonic acid tert-butyl ethyl ester, tert-butyl ethyl propane-1,3-dioate, zlchem 692, acmc-1cjju, 1-tert-butyl ethyl malonate
PubChem CID 96345
IUPAC Name 3-O-tert-butyl 1-O-ethyl propanedioate
SMILES CCOC(=O)CC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1192714
View Documents
Thermo Scientific Chemicals
A1192714
25 g
Each for $300.10
Only null left
 
AAA1192722
View Documents
Thermo Scientific Chemicals
A1192722
100 g
Each for $910.90
Only null left
 
Description

Description

tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.

Solubility
Miscible with most common solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

32864-38-3
188.223
OCOBFMZGRJOSOU-UHFFFAOYSA-N
96345
CCOC(=O)CC(=O)OC(C)(C)C
C9H16O4
MFCD00009193
tert-butyl ethyl malonate, t-butyl ethyl malonate, propanedioic acid, 1,1-dimethylethyl ethyl ester, 1-tert-butyl 3-ethyl propanedioate, tert-butyl ethyl propanedioate, malonic acid tert-butyl ethyl ester, tert-butyl ethyl propane-1,3-dioate, zlchem 692, acmc-1cjju, 1-tert-butyl ethyl malonate
3-O-tert-butyl 1-O-ethyl propanedioate

Specifications

32864-38-3
98°C to 100°C (22 mmHg)
C9H16O4
MFCD00009193
1776681
Miscible with most common solvents. Immiscible with water.
CCOC(=O)CC(=O)OC(C)(C)C
188.223
188.22
tert-Butyl ethyl malonate
0.994
87°C (188°F)
1.416
25 g
tert-butyl ethyl malonate, t-butyl ethyl malonate, propanedioic acid, 1,1-dimethylethyl ethyl ester, 1-tert-butyl 3-ethyl propanedioate, tert-butyl ethyl propanedioate, malonic acid tert-butyl ethyl ester, tert-butyl ethyl propane-1,3-dioate, zlchem 692, acmc-1cjju, 1-tert-butyl ethyl malonate
OCOBFMZGRJOSOU-UHFFFAOYSA-N
3-O-tert-butyl 1-O-ethyl propanedioate
96345
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 251-269-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only