missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl glycolate, 94%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H35711MD

 View more versions of this product

Catalog No. AAH35711MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butyl glycolate
Colorless to Yellow
HOCH2CO2C(CH3)3
250 mg
WINGEFIITRDOLJ-UHFFFAOYSA-N
tert-butyl 2-hydroxyacetate
11815394
94%
50595-15-8
C6H12O3
MFCD03452739
tert-butyl glycolate, 2-tert-butyl glycolate, 2-t-butyl glycolate, glycolic acid tert-butyl ester, hydroxyacetic acid tert-butyl ester, acetic acid, hydroxy-, 1,1-dimethylethyl ester, tert-butyl hydroxyacetate, t-butyl 2-hydroxyacetate, tert-butyl2-hydroxyacetate
CC(C)(C)OC(=O)CO
132.16
132.16
Low Melting Solid

Chemical Identifiers

50595-15-8
132.16
WINGEFIITRDOLJ-UHFFFAOYSA-N
11815394
CC(C)(C)OC(=O)CO
C6H12O3
MFCD03452739
tert-butyl glycolate, 2-tert-butyl glycolate, 2-t-butyl glycolate, glycolic acid tert-butyl ester, hydroxyacetic acid tert-butyl ester, acetic acid, hydroxy-, 1,1-dimethylethyl ester, tert-butyl hydroxyacetate, t-butyl 2-hydroxyacetate, tert-butyl2-hydroxyacetate
tert-butyl 2-hydroxyacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only