Learn More
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
$35.02 - $84.35
Chemical Identifiers
CAS | 75-91-2 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.12 |
InChI Key | CIHOLLKRGTVIJN-UHFFFAOYSA-N |
Synonym | tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid |
PubChem CID | 6410 |
ChEBI | CHEBI:64090 |
IUPAC Name | 2-hydroperoxy-2-methylpropane |
SMILES | CC(C)(C)OO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC180340050
|
Thermo Scientific Chemicals
180340050 |
5 g | Plastic bottle |
Each for $35.02
|
|
||||
AC180342500
|
Thermo Scientific Chemicals
180342500 |
250 g | Plastic bottle |
Each for $77.07
|
|
||||
AC180345000
|
Thermo Scientific Chemicals
180345000 |
500 g | Plastic bottle |
Each for $84.35
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-91-2 | |
90.12 | |
tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid | |
CHEBI:64090 | |
CC(C)(C)OO |
C4H10O2 | |
CIHOLLKRGTVIJN-UHFFFAOYSA-N | |
6410 | |
2-hydroperoxy-2-methylpropane |
Specifications
75-91-2,7732-18-5 | |
Colorless | |
37.0°C (15.0 mmHg) | |
Plastic bottle | |
1.3860 to 1.3880 | |
5 g | |
01,88; 02,49; 03,37; 05,75; 06,81; 07,43; 08,62; 09,78; 10,64; 11,91; 12,88; 14,60; 15,55; 16,53; 17,56 | |
15,1573 | |
Solubility in water: miscible. Other solubilities: soluble in hydroxide solutions,soluble in most common organic solvents | |
CC(C)(C)OO | |
90.12 | |
4.1 mPa.s (20°C) | |
90.12 | |
tert-Butyl hydroperoxide |
-2.8°C | |
0.9400g/mL | |
43°C | |
C4H10O2 | |
(CH3)3COOH | |
01,IV,1616 | |
0.94 | |
tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid | |
CIHOLLKRGTVIJN-UHFFFAOYSA-N | |
2-hydroperoxy-2-methylpropane | |
6410 | |
CHEBI:64090 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 200-915-7
RUO – Research Use Only