Learn More
tert-Butyl isocyanide, 98%, Thermo Scientific Chemicals
$92.71 - $277.19
Identifiants chimiques
CAS | 7188-38-7 |
---|---|
Molecular Formula | C5H9N |
Molecular Weight (g/mol) | 83.134 |
MDL Number | MFCD00000002 |
InChI Key | FAGLEPBREOXSAC-UHFFFAOYSA-N |
Synonym | tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane |
PubChem CID | 23577 |
IUPAC Name | 2-isocyano-2-methylpropane |
SMILES | CC(C)(C)[N+]#[C-] |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
AAL1974703
|
Thermo Scientific Chemicals
L1974703 |
1 g |
chaque for $92.71
|
|
|||||
AAL1974706
|
Thermo Scientific Chemicals
L1974706 |
5 g |
chaque for $277.19
|
|
|||||
Description
tert-Butyl isocyanide is used in a tin-free procedure for alkyl radical reactions in the presence of a free-radical initiator. It is a useful intermediate in multicomponent reactions. It is also used in the synthesis of coumarines, 4H-chromenes, isoxazolines and to trap 2-cyclopropylidene-1,3-diones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl isocyanide is used in a tin-free procedure for alkyl radical reactions in the presence of a free-radical initiator. It is a useful intermediate in multicomponent reactions. It is also used in the synthesis of coumarines, 4H-chromenes, isoxazolines and to trap 2-cyclopropylidene-1,3-diones.
Solubility
Soluble in organic solvents such as ethanol, methanol, ether, toluene and dichloromethane.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Identifiants chimiques
7188-38-7 | |
83.134 | |
FAGLEPBREOXSAC-UHFFFAOYSA-N | |
23577 | |
CC(C)(C)[N+]#[C-] |
C5H9N | |
MFCD00000002 | |
tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane | |
2-isocyano-2-methylpropane |
Spécifications
7188-38-7 | |
90°C to 92°C | |
Stench | |
1.3755 | |
1 g | |
1903156 | |
Soluble in organic solvents such as ethanol,methanol,ether,toluene and dichloromethane. | |
CC(C)(C)[N+]#[C-] | |
83.134 | |
83.13 | |
tert-Butyl isocyanide |
0.735 | |
−2°C (28°F) | |
C5H9N | |
MFCD00000002 | |
UN1992 | |
tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane | |
FAGLEPBREOXSAC-UHFFFAOYSA-N | |
2-isocyano-2-methylpropane | |
23577 | |
98% |
Sécurité et manipulation
GHS H Statement
H225-H330
Highly flammable liquid and vapor.
Fatal if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P284-P303+P361+P353-P304+P340-P310-P370+P378q-P501c
H225-H330
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 230-553-5
missing translation for 'rtecsNumber' : EQ7102500
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only