missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl Isocyanide 95.0+%, TCI America™

Supplier:  TCI America B12745ML

 View more versions of this product

Catalog No. B12745ML


Explore available promotions

Description

Description

Specifications

Specifications

tert-Butyl Isocyanide
Colorless
C5H9N
5 mL
tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
CC(C)(C)[N+]#[C-]
83.134
83.13
Liquid
7188-38-7
92°C
MFCD00000002
1992
FAGLEPBREOXSAC-UHFFFAOYSA-N
2-isocyano-2-methylpropane
23577
≥95.0% (GC)

Chemical Identifiers

7188-38-7
83.134
FAGLEPBREOXSAC-UHFFFAOYSA-N
23577
CC(C)(C)[N+]#[C-]
C5H9N
MFCD00000002
tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
2-isocyano-2-methylpropane
Videos
Safety and Handling

Safety and Handling

RTECSNumber : EQ7102500

TSCA : No

SDS
Documents

Documents

Promotions

Promotions