missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Butyl L-Pyroglutamate 98.0+%, TCI America™
$31.52 - $96.22
Chemical Identifiers
CAS | 35418-16-7 |
---|---|
Molecular Formula | C9H15NO3 |
Molecular Weight (g/mol) | 185.223 |
MDL Number | MFCD06659481 |
InChI Key | QXGSPAGZWRTTOT-LURJTMIESA-N |
Synonym | s-tert-butyl 5-oxopyrrolidine-2-carboxylate, s-2-pyrrolidone-5-carboxylic acid t-butyl ester, tert-butyl 5-oxo-l-prolinate, l-pyroglutamic acid tert-butyl ester, tert-butyl s-2-pyrrolidone-5-carboxylate, tert-butyl l-pyroglutamate, tert-butyl 2s-5-oxopyrrolidine-2-carboxylate, s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester, l-pyroglutamicacidtert-butylester, tert-butyl s-5-oxopyrrolidine-2-carboxylate |
PubChem CID | 10888648 |
IUPAC Name | tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate |
SMILES | CC(C)(C)OC(=O)C1CCC(=O)N1 |
Chemical Identifiers
35418-16-7 | |
185.223 | |
QXGSPAGZWRTTOT-LURJTMIESA-N | |
10888648 | |
CC(C)(C)OC(=O)C1CCC(=O)N1 |
C9H15NO3 | |
MFCD06659481 | |
s-tert-butyl 5-oxopyrrolidine-2-carboxylate, s-2-pyrrolidone-5-carboxylic acid t-butyl ester, tert-butyl 5-oxo-l-prolinate, l-pyroglutamic acid tert-butyl ester, tert-butyl s-2-pyrrolidone-5-carboxylate, tert-butyl l-pyroglutamate, tert-butyl 2s-5-oxopyrrolidine-2-carboxylate, s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester, l-pyroglutamicacidtert-butylester, tert-butyl s-5-oxopyrrolidine-2-carboxylate | |
tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate |
Specifications
35418-16-7 | |
White | |
MFCD06659481 | |
s-tert-butyl 5-oxopyrrolidine-2-carboxylate, s-2-pyrrolidone-5-carboxylic acid t-butyl ester, tert-butyl 5-oxo-l-prolinate, l-pyroglutamic acid tert-butyl ester, tert-butyl s-2-pyrrolidone-5-carboxylate, tert-butyl l-pyroglutamate, tert-butyl 2s-5-oxopyrrolidine-2-carboxylate, s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester, l-pyroglutamicacidtert-butylester, tert-butyl s-5-oxopyrrolidine-2-carboxylate | |
CC(C)(C)OC(=O)C1CCC(=O)N1 | |
185.223 | |
185.22 | |
Crystalline Powder |
104°C | |
C9H15NO3 | |
1 g | |
QXGSPAGZWRTTOT-LURJTMIESA-N | |
tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate | |
10888648 | |
≥98.0% (GC) | |
tert-Butyl L-Pyroglutamate |
Safety and Handling
TSCA : No