Learn More
tert-Butyl methyl ether, for HPLC
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
$223.74 - $223.74
Chemical Identifiers
| CAS | 1634-04-4 |
|---|---|
| Molecular Formula | C5H12O |
| Molecular Weight (g/mol) | 88.15 |
| MDL Number | MFCD00008812 |
| InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
| Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
| PubChem CID | 15413 |
| ChEBI | CHEBI:27642 |
| IUPAC Name | 2-methoxy-2-methylpropane |
| SMILES | CC(C)(C)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389050010
|
Thermo Scientific Chemicals
389050010 |
1 L | Glass Bottle |
Each for $223.74
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCChemical Identifiers
| 1634-04-4 | |
| 88.15 | |
| BZLVMXJERCGZMT-UHFFFAOYSA-N | |
| 15413 | |
| 2-methoxy-2-methylpropane |
| C5H12O | |
| MFCD00008812 | |
| tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
| CHEBI:27642 | |
| CC(C)(C)OC |
Specifications
| -110.0°C | |
| (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.009 max. at 280nm, 0.097 max. at 255nm, 0.301 max. at 240nm | |
| 0.0005 meq/g max. | |
| 0.7404g/mL | |
| 1 L | |
| Glass Bottle | |
| 1.3675 to 1.3695 | |
| 0.7404 | |
| (CH3)3COCH3 | |
| 0.36 mPa.s (20°C) | |
| 15, 6106 | |
| Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g | |
| CC(C)(C)OC | |
| 88.15 | |
| CHEBI:27642 | |
| ≥99.8% | |
| tert-Butyl methyl ether |
| 1634-04-4 | |
| Colorless | |
| 0.0002 meq/g max. | |
| 54.0°C to 56.0°C | |
| −28°C | |
| Liquid | |
| 0.0003% max. | |
| C5H12O | |
| MFCD00008812 | |
| 0.05% max. | |
| tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
| BZLVMXJERCGZMT-UHFFFAOYSA-N | |
| 2-methoxy-2-methylpropane | |
| 15413 | |
| 88.15 | |
| HPLC |
Safety and Handling
GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only