missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 366350050

 View more versions of this product

Catalog No. AC366350050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Authentic
Glass bottle
MFCD02179439
4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
HSJNIOYPTSKQBD-UHFFFAOYSA-N
tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
2734617
97+%
330793-01-6
97% min. (HPLC)
C17H26BNO4
5g
Solubility in water: insoluble in water
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)OC(C)(C)C
319.21
319.21

Chemical Identifiers

330793-01-6
319.21
HSJNIOYPTSKQBD-UHFFFAOYSA-N
2734617
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)OC(C)(C)C
C17H26BNO4
MFCD02179439
4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only