missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl N-[4-(aminomethyl)phenyl]carbamate, 97%, Thermo Scientific™

$277.24 - $1,265.67

Chemical Identifiers

CAS 220298-96-4
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD02183573
InChI Key URXUHALBOWYXJZ-UHFFFAOYSA-N
Synonym tert-butyl n-4-aminomethyl phenyl carbamate, 4-aminomethyl-1-n-boc-aniline, tert-butyl 4-aminomethyl phenyl carbamate, 4-boc-amino benzylamine, 4-aminomethy-1-n-boc-aniline, 4-n-boc-amino benzylamine, 4-tert-butoxycarbonylamino benzylamine, 4-aminomethyl-n-boc-aniline, tert-butyl-n-4-aminomethyl phenyl carbamate
PubChem CID 2794659
IUPAC Name tert-butyl N-[4-(aminomethyl)phenyl]carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MO01028DA
SDS View Documents
Thermo Scientific Chemicals
MO01028DA
1 g N/A
 
MO01028EA
SDS View Documents
Thermo Scientific Chemicals
MO01028EA
10 g N/A
 
Description

Description

Specifications

Chemical Identifiers

220298-96-4
222.29
URXUHALBOWYXJZ-UHFFFAOYSA-N
2794659
CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1
C12H18N2O2
MFCD02183573
tert-butyl n-4-aminomethyl phenyl carbamate, 4-aminomethyl-1-n-boc-aniline, tert-butyl 4-aminomethyl phenyl carbamate, 4-boc-amino benzylamine, 4-aminomethy-1-n-boc-aniline, 4-n-boc-amino benzylamine, 4-tert-butoxycarbonylamino benzylamine, 4-aminomethyl-n-boc-aniline, tert-butyl-n-4-aminomethyl phenyl carbamate
tert-butyl N-[4-(aminomethyl)phenyl]carbamate

Specifications

220298-96-4
95%
C12H18N2O2
1 g
URXUHALBOWYXJZ-UHFFFAOYSA-N
tert-butyl N-[4-(aminomethyl)phenyl]carbamate
2794659
97%
97.0
Amber Glass Bottle
MFCD02183573
tert-butyl n-4-aminomethyl phenyl carbamate, 4-aminomethyl-1-n-boc-aniline, tert-butyl 4-aminomethyl phenyl carbamate, 4-boc-amino benzylamine, 4-aminomethy-1-n-boc-aniline, 4-n-boc-amino benzylamine, 4-tert-butoxycarbonylamino benzylamine, 4-aminomethyl-n-boc-aniline, tert-butyl-n-4-aminomethyl phenyl carbamate
CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1
222.29
222.29
tert-Butyl N-[4-(aminomethyl)phenyl]carbamate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'