missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl N-(Benzyloxy)carbamate 98.0+%, TCI America™
$154.71 - $452.97
Chemical Identifiers
| CAS | 79722-21-7 |
|---|---|
| Molecular Formula | C12H17NO3 |
| Molecular Weight (g/mol) | 223.272 |
| MDL Number | MFCD00191873 |
| InChI Key | MZNBNPWFHGWAGH-UHFFFAOYSA-N |
| Synonym | N-(Benzyloxy)carbamic Acid tert-Butyl Ester |
| PubChem CID | 736159 |
| IUPAC Name | tert-butyl N-phenylmethoxycarbamate |
| SMILES | CC(C)(C)OC(=O)NOCC1=CC=CC=C1 |
Chemical Identifiers
| 79722-21-7 | |
| 223.272 | |
| MZNBNPWFHGWAGH-UHFFFAOYSA-N | |
| 736159 | |
| CC(C)(C)OC(=O)NOCC1=CC=CC=C1 |
| C12H17NO3 | |
| MFCD00191873 | |
| N-(Benzyloxy)carbamic Acid tert-Butyl Ester | |
| tert-butyl N-phenylmethoxycarbamate |
Specifications
| 79722-21-7 | |
| White-Yellow | |
| MFCD00191873 | |
| N-(Benzyloxy)carbamic Acid tert-Butyl Ester | |
| CC(C)(C)OC(=O)NOCC1=CC=CC=C1 | |
| 223.272 | |
| 223.27 | |
| Crystalline Powder |
| 48°C | |
| C12H17NO3 | |
| 5 g | |
| MZNBNPWFHGWAGH-UHFFFAOYSA-N | |
| tert-butyl N-phenylmethoxycarbamate | |
| 736159 | |
| ≥98.0% (N) | |
| tert-Butyl N-(Benzyloxy)carbamate |
Safety and Handling
TSCA : No