missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl nitrite, 90%, pure
CAS: 540-80-7 | C4H9NO2 | 103.12 g/mol
$161.58 - $757.33
Chemical Identifiers
| CAS | 540-80-7 |
|---|---|
| InChI Key | IOGXOCVLYRDXLW-UHFFFAOYSA-N |
| Synonym | t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono |
| PubChem CID | 10906 |
| IUPAC Name | tert-butyl nitrite |
| SMILES | CC(C)(C)ON=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC216221000
|
Thermo Scientific Chemicals
216221000 |
100 g | Glass bottle |
Each for $161.58
|
|
||||
|
AC216225000
|
Thermo Scientific Chemicals
216225000 |
500 g | Glass bottle |
Each for $757.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 540-80-7 | |
| t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono | |
| tert-butyl nitrite |
| IOGXOCVLYRDXLW-UHFFFAOYSA-N | |
| 10906 | |
| CC(C)(C)ON=O |
Specifications
| 540-80-7 | |
| 0.8600g/mL | |
| −10°C | |
| 88% min. (GC) | |
| C4H9NO2 | |
| (CH3)3CNO2 | |
| 01,382 | |
| 0.86 | |
| t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono | |
| IOGXOCVLYRDXLW-UHFFFAOYSA-N | |
| tert-butyl nitrite | |
| 10906 | |
| 90% | |
| Liquid |
| Yellow | |
| 61.0°C to 63.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3670 to 1.3700 | |
| 100 g | |
| 07,48 | |
| 15,1586 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,chloroform and carbon,disulfide,practically insoluble in glycerol | |
| CC(C)(C)ON=O | |
| 103.12 | |
| 103.12 | |
| Pure | |
| tert-Butyl nitrite |
Safety and Handling
Danger
RUO – Research Use Only