Learn More
tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate, 95%, Thermo Scientific Chemicals
$114.51 - $436.45
Chemical Identifiers
CAS | 35418-16-7 |
---|---|
Molecular Formula | C9H15NO3 |
Molecular Weight (g/mol) | 185.223 |
MDL Number | MFCD06659481 |
InChI Key | QXGSPAGZWRTTOT-LURJTMIESA-N |
Synonym | s-tert-butyl 5-oxopyrrolidine-2-carboxylate, s-2-pyrrolidone-5-carboxylic acid t-butyl ester, tert-butyl 5-oxo-l-prolinate, l-pyroglutamic acid tert-butyl ester, tert-butyl s-2-pyrrolidone-5-carboxylate, tert-butyl l-pyroglutamate, tert-butyl 2s-5-oxopyrrolidine-2-carboxylate, s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester, l-pyroglutamicacidtert-butylester, tert-butyl s-5-oxopyrrolidine-2-carboxylate |
PubChem CID | 10888648 |
IUPAC Name | tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate |
SMILES | CC(C)(C)OC(=O)C1CCC(=O)N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6538506
|
Thermo Scientific Chemicals
H6538506 |
5 g |
Each for $114.51
|
|
||||||
AAH6538514
|
Thermo Scientific Chemicals
H6538514 |
25 g |
Each for $436.45
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35418-16-7 | |
185.223 | |
QXGSPAGZWRTTOT-LURJTMIESA-N | |
10888648 | |
CC(C)(C)OC(=O)C1CCC(=O)N1 |
C9H15NO3 | |
MFCD06659481 | |
s-tert-butyl 5-oxopyrrolidine-2-carboxylate, s-2-pyrrolidone-5-carboxylic acid t-butyl ester, tert-butyl 5-oxo-l-prolinate, l-pyroglutamic acid tert-butyl ester, tert-butyl s-2-pyrrolidone-5-carboxylate, tert-butyl l-pyroglutamate, tert-butyl 2s-5-oxopyrrolidine-2-carboxylate, s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester, l-pyroglutamicacidtert-butylester, tert-butyl s-5-oxopyrrolidine-2-carboxylate | |
tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate |
Specifications
35418-16-7 | |
C9H15NO3 | |
5 g | |
s-tert-butyl 5-oxopyrrolidine-2-carboxylate, s-2-pyrrolidone-5-carboxylic acid t-butyl ester, tert-butyl 5-oxo-l-prolinate, l-pyroglutamic acid tert-butyl ester, tert-butyl s-2-pyrrolidone-5-carboxylate, tert-butyl l-pyroglutamate, tert-butyl 2s-5-oxopyrrolidine-2-carboxylate, s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester, l-pyroglutamicacidtert-butylester, tert-butyl s-5-oxopyrrolidine-2-carboxylate | |
CC(C)(C)OC(=O)C1CCC(=O)N1 | |
185.223 | |
185.22 | |
Powder |
White | |
MFCD06659481 | |
3590226 | |
QXGSPAGZWRTTOT-LURJTMIESA-N | |
tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate | |
10888648 | |
95% | |
tert-Butyl (S)-5-oxopyrrolidine-2-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 252-555-5
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only