Learn More
tert-Butyl vinyl ether, 98%, stab. with ca 0.1% N,N-diethylaniline, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3077014
Description
tert-Butyl vinyl ether is used in the synthesis of 3-tert-Butoxycyclobutanone by reacting with ketene using zinc(II) chloride as a catalyst. It acts as an intermediate for the functional vinyl monomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl vinyl ether is used in the synthesis of 3-tert-Butoxycyclobutanone by reacting with ketene using zinc(II) chloride as a catalyst. It acts as an intermediate for the functional vinyl monomer.
Solubility
Miscible with alcohol, carbon disulfide, chloroform and diethyl ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
tert-Butyl vinyl ether | |
926-02-3 | |
75°C to 76°C | |
Characteristic | |
1.399 | |
MFCD00048246 | |
UN3271 | |
Miscible with alcohol,carbon disulfide,chloroform and diethyl ether. Slightly miscible with water. | |
CC(C)(C)OC=C | |
100.16 | |
100.16 |
stabilized with ≈0.1% N,N-diethylaniline | |
0.762 | |
−17°C (1°F) | |
C6H12O | |
(CH3)3COCH=CH2 | |
25 g | |
tert-butyl vinyl ether, 2-ethenyloxy-2-methylpropane, propane, 2-ethenyloxy-2-methyl, propane,2-ethenyloxy-2-methyl, t-butyl vinyl ether, tert-butylvinyl ether, vinyltertiary-butylether, tertiarybutyl vinyl ether, vinyl tert.-butyl ether, tertiary butyl vinyl ether | |
PGYJSURPYAAOMM-UHFFFAOYSA-N | |
2-ethenoxy-2-methylpropane | |
70220 | |
98% |
Chemical Identifiers
926-02-3 | |
100.16 | |
PGYJSURPYAAOMM-UHFFFAOYSA-N | |
70220 | |
CC(C)(C)OC=C |
C6H12O | |
MFCD00048246 | |
tert-butyl vinyl ether, 2-ethenyloxy-2-methylpropane, propane, 2-ethenyloxy-2-methyl, propane,2-ethenyloxy-2-methyl, t-butyl vinyl ether, tert-butylvinyl ether, vinyltertiary-butylether, tertiarybutyl vinyl ether, vinyl tert.-butyl ether, tertiary butyl vinyl ether | |
2-ethenoxy-2-methylpropane |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H225-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 618-860-6
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only