missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butylacetyl chloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107790100

 View more versions of this product

Catalog No. AC107790100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butylacetyl chloride
7065-46-5
0.9600g/mL
21°C
99%
C6H11ClO
(CH3)3CCH2COCl
10 mL
3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
BUTKIHRNYUEGKB-UHFFFAOYSA-N
3,3-dimethylbutanoyl chloride
81514
99%
99%
Colorless to Yellow
127.0°C to 129.0°C
Authentic
Glass bottle
1.4210 to 1.4230
MFCD00000737
0.96
Solubility in water: reacts. Other solubilities: soluble in ethanol
CC(C)(C)CC(=O)Cl
134.61
134.61
Liquid

Chemical Identifiers

7065-46-5
134.61
BUTKIHRNYUEGKB-UHFFFAOYSA-N
81514
CC(C)(C)CC(=O)Cl
C6H11ClO
MFCD00000737
3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
3,3-dimethylbutanoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 230-356-4

Recommended Storage : Refrigerator +4°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only