missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Butylamine Hydroiodide 97.0+%, TCI America™
$193.56 - $663.60
Chemical Identifiers
CAS | 39557-45-4 |
---|---|
Molecular Formula | C4H12IN |
Molecular Weight (g/mol) | 201.051 |
InChI Key | NLJDBTZLVTWXRG-UHFFFAOYSA-N |
Synonym | tert-Butylammonium Iodide |
PubChem CID | 91972147 |
IUPAC Name | 2-methylpropan-2-amine;hydroiodide |
SMILES | CC(C)(C)N.I |
Chemical Identifiers
39557-45-4 | |
201.051 | |
tert-Butylammonium Iodide | |
2-methylpropan-2-amine;hydroiodide |
C4H12IN | |
NLJDBTZLVTWXRG-UHFFFAOYSA-N | |
91972147 | |
CC(C)(C)N.I |
Specifications
39557-45-4 | |
White | |
1 g | |
NLJDBTZLVTWXRG-UHFFFAOYSA-N | |
2-methylpropan-2-amine;hydroiodide | |
91972147 | |
≥97.0% (N,T) | |
tert-Butylamine Hydroiodide |
221°C | |
C4H12IN | |
tert-Butylammonium Iodide | |
CC(C)(C)N.I | |
201.051 | |
201.05 | |
Crystalline Powder |
Safety and Handling
TSCA : No