missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Butylamine 98.0+%, TCI America™
Supplier: TCI America B0709500ML
Specifications
tert-Butylamine | |
Colorless | |
C4H11N | |
500 mL | |
tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert | |
CC(C)(C)N | |
73.139 | |
CHEBI:44639 | |
≥98.0% (GC,T) |
75-64-9 | |
46°C | |
MFCD00008050 | |
3286 | |
YBRBMKDOPFTVDT-UHFFFAOYSA-N | |
2-methylpropan-2-amine | |
6385 | |
73.14 | |
Liquid |
Chemical Identifiers
75-64-9 | |
73.139 | |
YBRBMKDOPFTVDT-UHFFFAOYSA-N | |
6385 | |
2-methylpropan-2-amine |
C4H11N | |
MFCD00008050 | |
tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert | |
CHEBI:44639 | |
CC(C)(C)N |
Safety and Handling
EINECSNumber : (2)-0132
RTECSNumber : EO3330000
TSCA : Yes