missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butylchlorodimethylsilane, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 369101000

Catalog No. AC369101000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

tert-Butylchlorodimethylsilane
Colorless
58°C
C6H15ClSi
100 mL
15, 1567
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
tert-butyl-chloro-dimethylsilane
28928
150.72
18162-48-6,75-09-2
1.2250g/mL
AcroSeal™ Glass bottle
MFCD00000501
1.225
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
CC(C)(C)[Si](C)(C)Cl
150.72
CHEBI:85071
Liquid

Chemical Identifiers

18162-48-6
150.72
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
28928
tert-butyl-chloro-dimethylsilane
C6H15ClSi
MFCD00000501
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
CHEBI:85071
CC(C)(C)[Si](C)(C)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 242-042-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only