Learn More
tert-Butylcyclohexane, 99+%
CAS: 3178-22-1 | C10H20 | 140.27 g/mol
$99.58 - $326.58
Chemical Identifiers
| CAS | 3178-22-1 |
|---|---|
| Molecular Formula | C10H20 |
| Molecular Weight (g/mol) | 140.27 |
| MDL Number | MFCD00001471 |
| InChI Key | XTVMZZBLCLWBPM-UHFFFAOYSA-N |
| Synonym | t-butylcyclohexane, cyclohexane, tert-butyl, 1-tert-butylcyclohexane, cyclohexane, 1,1-dimethylethyl, unii-0rgt19ucq6, 1,1-dimethylethyl cyclohexane, 0rgt19ucq6, t-butyl cyclohexane, 4-t-butylcyclohexane, tert-butyl-cyclohexane |
| PubChem CID | 18508 |
| IUPAC Name | tert-butylcyclohexane |
| SMILES | CC(C)(C)C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2412614
|
Thermo Scientific Chemicals
B2412614 |
25 g |
N/A
|
|
|||||
|
AAB2412622
|
Thermo Scientific Chemicals
B2412622 |
100 g |
N/A
|
|
|||||
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 3178-22-1 | |
| 140.27 | |
| XTVMZZBLCLWBPM-UHFFFAOYSA-N | |
| 18508 | |
| CC(C)(C)C1CCCCC1 |
| C10H20 | |
| MFCD00001471 | |
| t-butylcyclohexane, cyclohexane, tert-butyl, 1-tert-butylcyclohexane, cyclohexane, 1,1-dimethylethyl, unii-0rgt19ucq6, 1,1-dimethylethyl cyclohexane, 0rgt19ucq6, t-butyl cyclohexane, 4-t-butylcyclohexane, tert-butyl-cyclohexane | |
| tert-butylcyclohexane |
Specifications
| 3178-22-1 | |
| 0.83 | |
| 40°C (104°F) | |
| 1.447 | |
| 25 g | |
| t-butylcyclohexane, cyclohexane, tert-butyl, 1-tert-butylcyclohexane, cyclohexane, 1,1-dimethylethyl, unii-0rgt19ucq6, 1,1-dimethylethyl cyclohexane, 0rgt19ucq6, t-butyl cyclohexane, 4-t-butylcyclohexane, tert-butyl-cyclohexane | |
| XTVMZZBLCLWBPM-UHFFFAOYSA-N | |
| tert-butylcyclohexane | |
| 18508 | |
| ≥99% |
| -41°C | |
| 171°C | |
| C10H20 | |
| MFCD00001471 | |
| UN3295 | |
| Insoluble in water. | |
| CC(C)(C)C1CCCCC1 | |
| 140.27 | |
| 140.27 | |
| tert-Butylcyclohexane |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 221-652-4
RTECSNumber : GU9384375
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only