missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Hexadecyl Mercaptan 78.0+%, TCI America™
Supplier: TCI America H006925ML
Specifications
| tert-Hexadecyl Mercaptan | |
| Yellow | |
| MFCD00043106 | |
| tert-Hexadecanethiol | |
| CC(C)(C)CCCCCCCCCCCCS | |
| 258.51 | |
| 258.51 | |
| Liquid |
| 25360-09-2 | |
| C16H34S | |
| 25 mL | |
| KZIYAWAYDPPPIU-UHFFFAOYSA-N | |
| 13,13-dimethyltetradecane-1-thiol | |
| 545889 | |
| ≥78.0% (T) |
Chemical Identifiers
| 25360-09-2 | |
| 258.51 | |
| KZIYAWAYDPPPIU-UHFFFAOYSA-N | |
| 545889 | |
| CC(C)(C)CCCCCCCCCCCCS |
| C16H34S | |
| MFCD00043106 | |
| tert-Hexadecanethiol | |
| 13,13-dimethyltetradecane-1-thiol |
Safety and Handling
EINECSNumber : (2)-0464
TSCA : Yes