missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Nitrosobutane dimer, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0046203

 View more versions of this product

Catalog No. AAL0046203


Explore available promotions

Description

Description

tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.

Solubility
Soluble in 5% chloroform.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

tert-Nitrosobutane dimer
88°C to 92°C (sublimation)
MFCD00002065
Light sensitive
Soluble in 5% chloroform.
CC(C)(C)N=O.CC(C)(C)N=O
174.244
174.24
6841-96-9
C8H18N2O2
1 g
dimer, nitroso-t-butane, nitroso-tert-butane, tert-nitrosobutane dimer, 2-methyl-2-nitrosopropane, dimer, bis, 2-methyl-2-nitroso propane dimer, 1,1,1 inverted exclamation marka,1 inverted exclamation marka-tetramethylazoethane dioxide
PKKJRWITDTTZCL-UHFFFAOYSA-N
2-methyl-2-nitrosopropane
44134826
≥98%

Chemical Identifiers

6841-96-9
174.244
PKKJRWITDTTZCL-UHFFFAOYSA-N
44134826
CC(C)(C)N=O.CC(C)(C)N=O
C8H18N2O2
MFCD00002065
dimer, nitroso-t-butane, nitroso-tert-butane, tert-nitrosobutane dimer, 2-methyl-2-nitrosopropane, dimer, bis, 2-methyl-2-nitroso propane dimer, 1,1,1 inverted exclamation marka,1 inverted exclamation marka-tetramethylazoethane dioxide
2-methyl-2-nitrosopropane
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only