missing translation for 'onlineSavingsMsg'
Learn More

tert-Octylamine, 95%

CAS: 107-45-9 | C8H19N | 129.24 g/mol

$39.44 - $136.01

Chemical Identifiers

CAS 107-45-9
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.24
MDL Number MFCD00008053
InChI Key QIJIUJYANDSEKG-UHFFFAOYSA-N
Synonym tert-octylamine, 1,1,3,3-tetramethylbutylamine, 2-pentanamine, 2,4,4-trimethyl, t-octylamine, 2,4,4-trimethyl-2-pentylamine, butylamine, 1,1,3,3-tetramethyl, 1,1,3,3-tetramethylbutanamine, butylamine, bis 1,3-dimethyl, unii-s84lil883b, 2,4,4-trimethyl-2-pentanamine
PubChem CID 61017
IUPAC Name 2,4,4-trimethylpentan-2-amine
SMILES CC(C)(C)CC(C)(C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC129500050
SDS View Documents
Thermo Scientific Chemicals
129500050
5 g Glass bottle
Each for $39.44
Only null left
 
AC129505000
SDS View Documents
Thermo Scientific Chemicals
129505000
500 g Glass bottle
Each for $136.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-45-9
129.24
QIJIUJYANDSEKG-UHFFFAOYSA-N
61017
CC(C)(C)CC(C)(C)N
C8H19N
MFCD00008053
tert-octylamine, 1,1,3,3-tetramethylbutylamine, 2-pentanamine, 2,4,4-trimethyl, t-octylamine, 2,4,4-trimethyl-2-pentylamine, butylamine, 1,1,3,3-tetramethyl, 1,1,3,3-tetramethylbutanamine, butylamine, bis 1,3-dimethyl, unii-s84lil883b, 2,4,4-trimethyl-2-pentanamine
2,4,4-trimethylpentan-2-amine

Specifications

107-45-9
0.8050g/mL
32°C
94% min. (GC)
C8H19N
(CH3)3CCH2C(CH3)2NH2
5 g
0.805
Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents
CC(C)(C)CC(C)(C)N
129.24
129.24
Liquid
-67.0°C
137.0°C to 143.0°C
Authentic
Glass bottle
1.4235 to 1.4255
MFCD00008053
04, 198
tert-octylamine, 1,1,3,3-tetramethylbutylamine, 2-pentanamine, 2,4,4-trimethyl, t-octylamine, 2,4,4-trimethyl-2-pentylamine, butylamine, 1,1,3,3-tetramethyl, 1,1,3,3-tetramethylbutanamine, butylamine, bis 1,3-dimethyl, unii-s84lil883b, 2,4,4-trimethyl-2-pentanamine
QIJIUJYANDSEKG-UHFFFAOYSA-N
2,4,4-trimethylpentan-2-amine
61017
95%
tert-Octylamine, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-491-1

RTECSNumber : EO3850000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only