missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Pentylbenzene, 97%, Thermo Scientific Chemicals
$88.22 - $296.66
Chemical Identifiers
CAS | 2049-95-8 |
---|---|
Molecular Formula | C11H16 |
Molecular Weight (g/mol) | 148.249 |
MDL Number | MFCD00015187 |
InChI Key | QHTJSSMHBLGUHV-UHFFFAOYSA-N |
Synonym | tert-amylbenzene, tert-pentylbenzene, 2-methyl-2-phenylbutane, 1,1-dimethylpropyl benzene, benzene, 1,1-dimethylpropyl, benzene, tert-pentyl, 2-phenyl-2-methylbutane, t-amylbenzene, 2-methylbutan-2-yl benzene, tert-amyl benzene |
PubChem CID | 16295 |
IUPAC Name | 2-methylbutan-2-ylbenzene |
SMILES | CCC(C)(C)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0373109
|
Thermo Scientific Chemicals
L0373109 |
10 g |
Each for $88.22
|
|
|||||
AAL0373118
|
Thermo Scientific Chemicals
L0373118 |
50 g |
Each for $296.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2049-95-8 | |
148.249 | |
QHTJSSMHBLGUHV-UHFFFAOYSA-N | |
16295 | |
CCC(C)(C)C1=CC=CC=C1 |
C11H16 | |
MFCD00015187 | |
tert-amylbenzene, tert-pentylbenzene, 2-methyl-2-phenylbutane, 1,1-dimethylpropyl benzene, benzene, 1,1-dimethylpropyl, benzene, tert-pentyl, 2-phenyl-2-methylbutane, t-amylbenzene, 2-methylbutan-2-yl benzene, tert-amyl benzene | |
2-methylbutan-2-ylbenzene |
Specifications
2049-95-8 | |
188°C to 191°C | |
C11H16 | |
MFCD00015187 | |
2039053 | |
QHTJSSMHBLGUHV-UHFFFAOYSA-N | |
2-methylbutan-2-ylbenzene | |
16295 | |
97% |
0.87 | |
65°C (149°F) | |
1.495 | |
10 g | |
tert-amylbenzene, tert-pentylbenzene, 2-methyl-2-phenylbutane, 1,1-dimethylpropyl benzene, benzene, 1,1-dimethylpropyl, benzene, tert-pentyl, 2-phenyl-2-methylbutane, t-amylbenzene, 2-methylbutan-2-yl benzene, tert-amyl benzene | |
CCC(C)(C)C1=CC=CC=C1 | |
148.249 | |
148.25 | |
tert-Pentylbenzene |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 218-076-0
RTECSNumber : DA6720000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only