Terumo™ Surflo™ Winged Infusion Sets
Secure, safe and simple I.V. therapy permits continuous access to a vein with minimum discomfort
Supplier:  Terumo™ SV23BLK
 View more versions of this product
Catalog No. 22-258-091
Explore available promotions
Description
Description
  • Siliconized needle bonded to flexible wings
  • Soft tubing attached to a Luer fitting with threaded venting cap
  • Needles bonded to wings at slight downward angle—better conform to body contours and reduce trauma to tunica intima
  • Color-coded wings according to needle gauge and embossed with gauge size
  • Soft polyvinyl chloride tubing resists kinking
Order Info
22-254-038 and 22-254-039 have a latex cap covering the end of the Luer fitting. With these sets, solutions can be intermittently injected (with a hypodermic syringe) directly through the cap and into the tubing.
Specifications
Specifications
Ultra Thin Wall Needle
PVC
Sterile
0.5 in.
Light Blue
23 G
Luer Fitting
12 in.