Learn More
Terumo™ Surshield™ Safety Winged Blood Collection Sets
One-handed activation
Supplier: Terumo™ MNSVS25B30
Description
- For use with evacuated blood collection tubes only; not compatible with I.V. therapy applications
- Designed for an evacuated blood collection system, which provides closed, conventional, and safe blood collection for fast and easy sampling and analysis
- Non-latex plastic and tubing ensure allergy-free usage
- One-handed operation makes siliconized needle insertion quick and easy with the flexible wings allowing for a secure grip
- Soft, flexible interlocking wings readily conform to the body's contour
- The wing is angled for ease of positioning of the needle during venipuncture, for less risk of vessel damage or occlusion of the lumen
- Wings are color coded according to needle gauge and embossed with gauge size
- After use, press the plastic Surshield into place over the needle to prevent accidental needlestick injuries before discarding
Specifications
| Winged Collection Set | |
| Sterile | |
| 12 in. |
| 25 G | |
| 0.75 in. |