Terumo™ Syringes with SurGuard3™ 22G Safety Hypodermic Needles
With three ways to activate (finger, thumb and hard surface) and a broad range of sizes, SurGuard™3 is the industry's most adaptable safety needle.
$69.37 - $69.37
Specifications
Needle Gauge 22 G
Latex Free Yes
Product Type Syringe
Volume (Metric) 3 mL
Sterility Sterile
View More Specs
Products 1
Catalog Number Mfr. No. Length (English) Needle Price Quantity  
Catalog Number Mfr. No. Length (English) Needle Price Quantity  
22-272-122
View Documents
Terumo™
SG303L2225
1 in.
Pack of 100 Each for $69.37
Case of 4 Pack for $277.48
Description
Description
  • 3 modes of activation-thumb, finger, or hard surface
  • Separate finger/thumb grips with ridges and nonslip surfaces
  • Hub and needle locks for security post activation
  • Broad range of sizes
  • Firm injection feel
  • Sharpest needles among major brands tested; 10% sharper than similar needles from Becton, Dickinson and Company
  • More comfortable injections for patients
Specifications
Specifications
22 G
Syringe
Sterile
Yes
3 mL
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'