Fisherbrand™ Test Paper
Use these test papers for a variety of applications.
Supplier:  Fisherbrand™ 19012V100
 View more versions of this product
Catalog No. S67171
Explore available promotions
Description
Description
  • A foolproof pH test to determine if a solution is acidic
  • The Blue Litmus test paper does not come with a color chart as it cannot provide you with a defined pH value
  • The test strip will turn red if dipped into an acidic solution (pH <7)
  • It will remain unchanged in an alkaline solution
  • We have tested the blue litmus with multiple pH buffers
  • The blue paper is blue at pH 8
  • At pH 7, the sample is maybe just a little red
  • The real change to red seems to start with pH 6
  • 5 and lower
  • Neutral Litmus test paper tests for Neutrality of a solution, or its acidity or alkalinity
  • The test strip will turn red if dipped into an acidic solution (pH <7) or blue if dipped into an alkaline solution (pH >7)
  • Red Litmus test paper is a foolproof pH test to determine if a solution is alkaline
  • The test strip will turn blue if dipped into an alkaline solution (pH >7)
  • Color chart not included
Specifications
Specifications
NFPA
For all laboratory moisture tests
Litmus Red Test Strip
7 to 14
Pk. of 12 Vials; 100 Strips/Vial