missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Supplier: Thermo Scientific™ 30183
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.Spécifications
Test Tube Rack | |
6 x 6 | |
14 to 16 mm Tubes, 15 mL Centrifuge Tubes | |
MaxQ™ Shaker Platform |
Orange; 14 to 16 mm; 6 x 6 array (fits 15 mL centrifuge tubes) | |
Orange | |
36 |