missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium fluoride trihydrate, 98%

CAS: 87749-50-6 | C16H42FNO3 | 315.514 g/mol

$130.27 - $319.52

Chemical Identifiers

CAS 87749-50-6
Molecular Formula C16H42FNO3
Molecular Weight (g/mol) 315.514
MDL Number MFCD00149981
InChI Key VEPTXBCIDSFGBF-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
PubChem CID 11726816
IUPAC Name tetrabutylazanium;fluoride;trihydrate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1330309
View Documents
Thermo Scientific Chemicals
L1330309
10 g
Each for $130.27
Only null left
 
AAL1330318
View Documents
Thermo Scientific Chemicals
L1330318
50 g
Each for $319.52
Only null left
 
Description

Description

Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

Solubility
Soluble in water, terahydrofuran, dimethyl sulfoxide and acetonitrile.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

87749-50-6
315.514
VEPTXBCIDSFGBF-UHFFFAOYSA-M
11726816
CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]
C16H42FNO3
MFCD00149981
tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
tetrabutylazanium;fluoride;trihydrate

Specifications

87749-50-6
Odorless
[CH3(CH2)3]4NF·3H2O
10 g
3761900
tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
VEPTXBCIDSFGBF-UHFFFAOYSA-M
tetrabutylazanium;fluoride;trihydrate
11726816
98%
54°C to 60°C
C16H42FNO3
MFCD00149981
UN2811
Hygroscopic
Soluble in water,terahydrofuran,dimethyl sulfoxide and acetonitrile.
CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]
315.514
315.52 (261.47 Anhydrous)
Tetra-n-butylammonium fluoride trihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H318-H315
Fatal if swallowed.
Causes serious eye damage.
Causes skin irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 207-057-2

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only