missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetra-n-butylammonium iodide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A154840B

 View more versions of this product

Catalog No. AAA154840B


Explore available promotions

Description

Description

Tetra-n-butylammonium iodide is used as a catalyst for methylation and benzylation of alcohols with dimethyl sulfate and benzyl bromide respectively. It is a useful source of iodide anion and involved in the preparation of neopentyl iodides from the triflates. It is also used in the preparation of novel quaternary amines, which serve as an antibacterial agent in the rise of drug-resistant bacteria. It acts as a phosphonium reversible inhibitors of cholinesterases. It is associated with boron trifluoride diethyl¬ etherate and used in the cleavage of ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium iodide is used as a catalyst for methylation and benzylation of alcohols with dimethyl sulfate and benzyl bromide respectively. It is a useful source of iodide anion and involved in the preparation of neopentyl iodides from the triflates. It is also used in the preparation of novel quaternary amines, which serve as an antibacterial agent in the rise of drug-resistant bacteria. It acts as a phosphonium reversible inhibitors of cholinesterases. It is associated with boron trifluoride diethyl¬ etherate and used in the cleavage of ethers.

Solubility
Soluble in water and methanol. Insoluble in benzene.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Tetra-n-butylammonium iodide
142°C to 148°C
MFCD00011636
3916152
tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
DPKBAXPHAYBPRL-UHFFFAOYSA-M
tetrabutylazanium;iodide
67553
98%
311-28-4
C16H36IN
1000 g
Hygroscopic; Light sensitive
Soluble in water and methanol. Insoluble in benzene.
[I-].CCCC[N+](CCCC)(CCCC)CCCC
369.38
369.38

Chemical Identifiers

311-28-4
369.38
DPKBAXPHAYBPRL-UHFFFAOYSA-M
67553
[I-].CCCC[N+](CCCC)(CCCC)CCCC
C16H36IN
MFCD00011636
tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
tetrabutylazanium;iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 206-220-5

RTECSNumber : BS5450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only