Learn More
Tetra-n-butylphosphonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
$97.77 - $1073.70
Chemical Identifiers
CAS | 14518-69-5 |
---|---|
Molecular Formula | C16H37OP |
Molecular Weight (g/mol) | 276.45 |
MDL Number | MFCD00068456 |
InChI Key | DFQPZDGUFQJANM-UHFFFAOYSA-M |
Synonym | tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o |
PubChem CID | 84487 |
IUPAC Name | tetrabutylphosphanium;hydroxide |
SMILES | [OH-].CCCC[P+](CCCC)(CCCC)CCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA41718AD
|
Thermo Scientific Chemicals
041718AD |
50 mL |
Each for $97.77
|
|
|||||
AA41718AK
|
Thermo Scientific Chemicals
041718AK |
250 mL |
Each for $371.59
|
|
|||||
AA41718K2
|
Thermo Scientific Chemicals
041718K2 |
1 L |
Each for $1,073.70
|
|
|||||
Description
Tetra-n-butylphosphonium hydroxide is used for the extraction of wheat straw and dissolution of wood. It is also used to produce hydroxide ion or proton conductive ionic liquids by mixing it with benzimidazole. It can be used as a phase transfer catalyst in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14518-69-5 | |
276.45 | |
DFQPZDGUFQJANM-UHFFFAOYSA-M | |
84487 | |
[OH-].CCCC[P+](CCCC)(CCCC)CCCC |
C16H37OP | |
MFCD00068456 | |
tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o | |
tetrabutylphosphanium;hydroxide |
Specifications
14518-69-5 | |
0.989 g/mL | |
1.412 | |
MFCD00068456 | |
UN3267 | |
Miscible with water. | |
[OH-].CCCC[P+](CCCC)(CCCC)CCCC | |
276.45 | |
276.45 | |
Liquid |
Colorless to Yellow | |
C16H37OP | |
[CH3(CH2)3]4POH | |
50 mL | |
tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o | |
DFQPZDGUFQJANM-UHFFFAOYSA-M | |
tetrabutylphosphanium;hydroxide | |
84487 | |
40% w/w aq. soln. | |
Tetra-n-butylphosphonium hydroxide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 238-528-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only